1-Methylpyrrol-2-carbonsäure

Lieferant: COMBI-BLOCKS

HI-1503-5G HI-1503-25G HI-1503-1G
COBBHI-1503-5GEA 0 EUR
COBBHI-1503-5G COBBHI-1503-25G COBBHI-1503-1G
1-Methylpyrrol-2-carbonsäure
1-Methylpyrrole-2-carboxylic acid
Formel: C₆H₇NO₂
MDL: MFCD00003088
CAS-Nummer: 6973-60-0
EINECS: 230-217-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR