1-Methyl-2-(trichloracetyl)pyrrol

Lieferant: BIONET RESEARCH

Synonyme: 2,2,2-Trichloro-1-(1-methyl-1H-pyrrol-2-yl)ethanone

5D-093.500 5D-093.1 5D-093.5 5D-093.10
BIOT5D-093.500EA 0 EUR
BIOT5D-093.500 BIOT5D-093.1 BIOT5D-093.5 BIOT5D-093.10
1-Methyl-2-(trichloracetyl)pyrrol
1-Methyl-2-(trichloracetyl)pyrrol
Formel: C₇H₆Cl₃NO
Molecular Weight: 226,4876 g/mol
MDL: MFCD00052521
CAS-Nummer: 21898-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR