4-Acetyl-2-(trichloracetyl)pyrrol

Lieferant: BIONET RESEARCH

Synonyme: 1-(4-Acetyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

9R-0288.500 9R-0288.1 9R-0288.5 9R-0288.10
BIOT9R-0288.500EA 0 EUR
BIOT9R-0288.500 BIOT9R-0288.1 BIOT9R-0288.5 BIOT9R-0288.10
4-Acetyl-2-(trichloracetyl)pyrrol
4-Acetyl-2-(trichloracetyl)pyrrol
Formel: C₈H₆Cl₃NO₂
MDL: MFCD03001304
CAS-Nummer: 72652-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR