3-(1-Pyrrolyl)benzonitril

Lieferant: Thermo Scientific

BTB02480.1 BTB02480.10 BTB02480.50
MAYBBTB02480.1EA 0 EUR
MAYBBTB02480.1 MAYBBTB02480.10 MAYBBTB02480.50
3-(1-Pyrrolyl)benzonitril
3-(1-Pyrrolyl)benzonitril
Formel: C₁₁H₈N₂
MDL: MFCD00052877
CAS-Nummer: 175134-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR