2-Fluor-6-(1-pyrrolyl)benzonitril
Lieferant: Thermo Scientific
CD05646.1
CD05646.10
MAYBCD05646.1EA
0
EUR
MAYBCD05646.1
MAYBCD05646.10
2-Fluor-6-(1-pyrrolyl)benzonitril
2-Fluor-6-(1-pyrrolyl)benzonitril
Formel:
C₁₁H₇FN₂ |
MDL:
MFCD00052222 CAS-Nummer: 148901-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...