Triglycidylisocyanurat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3,5-Triglycidyl isocyanurate

M-Clarity™ Qualitätsstufe = MQ200

379506-100G 379506-500G
SIAL379506-100GEA 114 EUR
SIAL379506-100G SIAL379506-500G
Triglycidylisocyanurat, Sigma-Aldrich®
Triglycidylisocyanurat
Formel: C₁₂H₁₅N₃O₆
Molecular Weight: 297,2640 g/mol
MDL: MFCD00080670
CAS-Nummer: 2451-62-9
EINECS: 219-514-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR