2-Amino-4-methoxy-6-(methylthio)-1,3,5-triazin

Lieferant: Thermo Scientific

RF03315.1 RF03315.10
MAYBRF03315.1EA 0 EUR
MAYBRF03315.1 MAYBRF03315.10
2-Amino-4-methoxy-6-(methylthio)-1,3,5-triazin
2-Amino-4-methoxy-6-(methylthio)-1,3,5-triazine
Formel: C₅H₈N₄OS
MDL: MFCD00052779
CAS-Nummer: 30358-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR