Diethoxymethylphenylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Diethoxy(methyl)(phenyl)silane

SILS11525.10 SILS11525.25 SILS11525.100
SILNSILS11525.10EA 0 EUR
SILNSILS11525.10 SILNSILS11525.25 SILNSILS11525.100
Diethoxymethylphenylsilan
Diethoxymethylphenylsilan
Formel: C₁₁H₁₈O₂Si
Molecular Weight: 210,35 g/mol
MDL: MFCD00048541
CAS-Nummer: 775-56-4
EINECS: 212-275-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR