(Chlormethyl)dimethylphenylsilan
Lieferant: CHEMPUR SILANE
Synonyme:
(Chloromethyl)dimethyl(phenyl)silane
SILS04050.10
SILS04050.25
SILS04050.100
SILNSILS04050.10EA
0
EUR
SILNSILS04050.10
SILNSILS04050.25
SILNSILS04050.100
(Chlormethyl)dimethylphenylsilan
(Chlormethyl)dimethylphenylsilan
Formel:
C₉H₁₃ClSi Molecular Weight: 184,7400 g/mol |
MDL:
MFCD00040858 CAS-Nummer: 1833-51-8 EINECS: 217-392-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...