1-Phenyl-1-(trimethylsilyloxy)ethylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trimethyl[(1-phenylvinyl)oxy]silane, 1-Phenyl-1-trimethylsiloxyethylene

M-Clarity™ Qualitätsstufe = MQ100

235040-25G 235040-5G
SIAL235040-25GEA 272 EUR
SIAL235040-25G SIAL235040-5G
1-Phenyl-1-(trimethylsilyloxy)ethylen, Sigma-Aldrich®
1-Phenyl-1-(trimethylsilyloxy)ethylen
Formel: C₁₁H₁₆OSi
Molecular Weight: 192,34 g/mol
MDL: MFCD00008582
CAS-Nummer: 13735-81-4
EINECS: 237-308-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR