1-Phenyl-1-(trimethylsilyloxy)ethylen

Lieferant: CHEMPUR SILANE

Synonyme: Trimethyl[(1-phenylvinyl)oxy]silane, 1-Phenyl-1-trimethylsiloxyethylene

SILS13776.5 SILS13776.25
SILNSILS13776.5EA 0 EUR
SILNSILS13776.5 SILNSILS13776.25
1-Phenyl-1-(trimethylsilyloxy)ethylen
1-Phenyl-1-(trimethylsilyloxy)ethylen
Formel: C₁₁H₁₆OSi
Molecular Weight: 192,34 g/mol
MDL: MFCD00008582
CAS-Nummer: 13735-81-4
EINECS: 237-308-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR