1,4-Bis(tributylstannyl)benzol, Sigma-Aldrich®

Lieferant: Merck
Gefahr

721204-1G 721204-5G
SIAL721204-1GEA 146 EUR
SIAL721204-1G SIAL721204-5G
1,4-Bis(tributylstannyl)benzol, Sigma-Aldrich®
1,4-Bis(tributylstannyl)benzol
Formel: C₃₀H₅₈Sn₂
MDL: MFCD17015284
CAS-Nummer: 17151-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR