4-Brom-2-piperazinopyridin

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-Bromo-2-pyridyl)piperazine

PY-1178-5G PY-1178-1G
COBBPY-1178-5GEA 1020 EUR
COBBPY-1178-5G COBBPY-1178-1G
4-Brom-2-piperazinopyridin
4-Brom-2-piperazinopyridin
Formel: C₉H₁₂BrN₃
Molecular Weight: 242,1157 g/mol
MDL: MFCD16618975
CAS-Nummer: 1201643-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR