Nicotinamidin Hydrochlorid 97%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 3-Amidinopyridinium chloride, 3-Amidinopyridine hydrochloride, Pyridine-3-carboximidamide Monohydrochloride, Pyridine-3-carboximidamide hydrochloride, 3-Carbamimidoylpyridine Monohydrochloride
B20406.03 B20406.06
B20406.03EA 23.3 EUR
B20406.03 B20406.06
Nicotinamidin Hydrochlorid 97%
Nicotinamidin Hydrochlorid
Purity: 97% CAS No.: 7356-60-7 Molecular Formula: C6H7N3.HCl Formula Weight: 157.60 Melting Point: 187-189° MDL No.: MFCD00054326
Formel: C₆H₈ClN₃
Molecular Weight: 157.60 g/mol
Schmelzpunkt: 187…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00054326
CAS-Nummer: 7356-60-7
Bestellen
TECHNISCHE DATEN
Environmentally Preferable