Nicotinamidin Hydrochlorid

Lieferant: Thermo Scientific

Synonyme: 3-Amidinopyridinium chloride, 3-Amidinopyridine hydrochloride, Pyridine-3-carboximidamide Monohydrochloride, Pyridine-3-carboximidamide hydrochloride, 3-Carbamimidoylpyridine Monohydrochloride

SEW03691.1 SEW03691.10
MAYBSEW03691.1EA 0 EUR
MAYBSEW03691.1 MAYBSEW03691.10
Nicotinamidin Hydrochlorid
Nicotinamidin Hydrochlorid
Formel: C₆H₈ClN₃
Molecular Weight: 157,60 g/mol
MDL: MFCD00054326
CAS-Nummer: 7356-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR