4-(5-Oxazolyl)benzonitril

Lieferant: Thermo Scientific

DFP00102.1 DFP00102.10
MAYBDFP00102.1EA 0 EUR
MAYBDFP00102.1 MAYBDFP00102.10
4-(5-Oxazolyl)benzonitril
4-(5-Oxazolyl)benzonitril
Formel: C₁₀H₆N₂O
Molecular Weight: 170,1674 g/mol
MDL: MFCD00084987
CAS-Nummer: 87150-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR