1-(2-Bromphenyl)-3-methylthioharnstoff

Lieferant: COMBI-BLOCKS

OT-0653-1G OT-0653-5G
COBBOT-0653-1GEA 204 EUR
COBBOT-0653-1G COBBOT-0653-5G
1-(2-Bromphenyl)-3-methylthioharnstoff
1-(2-Bromphenyl)-3-methylthioharnstoff
Formel: C₈H₉BrN₂S
MDL: MFCD06629750
CAS-Nummer: 52599-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR