1-(4-Brom-2-fluorphenyl)thioharnstoff

Lieferant: COMBI-BLOCKS

HC-3369-1G HC-3369-25G HC-3369-5G
COBBHC-3369-1GEA 204 EUR
COBBHC-3369-1G COBBHC-3369-25G COBBHC-3369-5G
1-(4-Brom-2-fluorphenyl)thioharnstoff
1-(4-Brom-2-fluorphenyl)thioharnstoff
Formel: C₇H₆BrFN₂S
MDL: MFCD09702294
CAS-Nummer: 930396-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR