N-Boc-D-alaninol 98%, ee 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-D-Alaninol, (R)-2-(BOC-amino)-1-propanol
H27887.03
H27887.06
H27887.14
H27887.03EA
44.4
EUR
H27887.03
H27887.06
H27887.14
N-Boc-D-alaninol 98%, ee 98%
N-Boc-D-alaninol
Purity: 98%
CAS No.: 106391-86-0
Molecular Formula: C8H17NO3
Formula Weight: 175.23
Melting Point: 58-62°
Optical Rotation: +11° (c=1 in chloroform)
Formel:
C₈H₁₇NO₃ Molecular Weight: 175.2 g/mol Schmelzpunkt: 58…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00235912 CAS-Nummer: 106391-86-0 |