3-Pyridyloxyessigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

B24780.03 B24780.06 B24780.14
B24780.03EA 47.2 EUR
B24780.03 B24780.06 B24780.14
3-Pyridyloxyessigsäure ≥98%
3-Pyridyloxyessigsäure

Purity: 98% CAS No.: 86649-57-2 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 210-212° MDL No.: MFCD04039772

Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 210…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039772
CAS-Nummer: 86649-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR