N-Acetyl-2-fluor-4-(3-hydroxy-3-methylbut-1-inyl)anilin
Lieferant: COMBI-BLOCKS
HC-3079-5G
HC-3079-1G
COBBHC-3079-5GEA
451
EUR
COBBHC-3079-5G
COBBHC-3079-1G
N-Acetyl-2-fluor-4-(3-hydroxy-3-methylbut-1-inyl)anilin
N-Acetyl-2-fluor-4-(3-hydroxy-3-methylbut-1-inyl)anilin
Formel:
C₁₃H₁₄FNO₂ |
MDL:
MFCD08701794 CAS-Nummer: 893642-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...