2-(Methylthio)ethylamin

Lieferant: Thermo Scientific

Synonyme: 2-(Methylthio)ethanamine

CD06838DA CD06838EA
MAYBCD06838DAEA 0 EUR
MAYBCD06838DA MAYBCD06838EA
2-(Methylthio)ethylamin
2-(Methylthio)ethylamin
Formel: C₃H₉NS
Molecular Weight: 91.18 g/mol
MDL: MFCD00014825
CAS-Nummer: 18542-42-2
EINECS: 242-412-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR