N-Boc-D-valin

Lieferant: BACHEM BIOCHEMICA

Synonyme: Boc-D-Val-OH

4000559.0005 4000559.0025
BACE4000559.0005EA 0 EUR
BACE4000559.0005 BACE4000559.0025
N-Boc-D-valin
N-Boc-D-valin
Formel: C₁₀H₁₉NO₄
Molecular Weight: 217.3 g/mol
MDL: MFCD00038282
CAS-Nummer: 22838-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR