N-Methyl-N-(2-pyridyl)pivalamid

Lieferant: COMBI-BLOCKS

Synonyme: N,2,2-Trimethyl-N-(pyridin-2-yl)propanamide

PY-7250-5G PY-7250-1G
COBBPY-7250-5GEA 1180 EUR
COBBPY-7250-5G COBBPY-7250-1G
N-Methyl-N-(2-pyridyl)pivalamid
N-Methyl-N-(2-pyridyl)pivalamid
Formel: C₁₁H₁₆N₂O
MDL: MFCD09878435
CAS-Nummer: 96830-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR