N-(3-Pyridyl)pivalamid
Lieferant: COMBI-BLOCKS
Synonyme:
2,2-Dimethyl-N-pyridin-3-yl-propionamide, 2,2-Dimehtyl-N-pyridin-3-yl-propionamide
HC-6925-5G
HC-6925-25G
HC-6925-1G
COBBHC-6925-5GEA
81.3
EUR
COBBHC-6925-5G
COBBHC-6925-25G
COBBHC-6925-1G
N-(3-Pyridyl)pivalamid
N-(3-Pyridyl)pivalamid
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178.23 g/mol |
MDL:
MFCD00996243 CAS-Nummer: 70298-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...