(2-Chlor-4-fluorphenyl)acetamid

Lieferant: Thermo Scientific

TL01011.1 TL01011.10
MAYBTL01011.1EA 0 EUR
MAYBTL01011.1 MAYBTL01011.10
(2-Chlor-4-fluorphenyl)acetamid
(2-Chlor-4-fluorphenyl)acetamid
Formel: C₈H₇ClFNO
MDL: MFCD02180958
CAS-Nummer: 306937-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR