N(α)-Benzyloxycarbonyl-D-lysin

Lieferant: BACHEM BIOCHEMICA

Synonyme: Cbz-D-Lys-OH, Z-D-Lys-OH

4010002.0001 4010002.0005
BACE4010002.0001EA 0 EUR
BACE4010002.0001 BACE4010002.0005
N(α)-Benzyloxycarbonyl-D-lysin
N(α)-Benzyloxycarbonyl-D-lysin
Formel: C₁₄H₂₀N₂O₄
Molecular Weight: 280,3 g/mol
MDL: MFCD00067713
CAS-Nummer: 70671-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR