3-(Boc-amino)-3-(4-methoxyphenyl)propionsäure
Lieferant: BIONET RESEARCH
Synonyme:
3-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid
9R-0657.500
9R-0657.1
BIOT9R-0657.500EA
0
EUR
BIOT9R-0657.500
BIOT9R-0657.1
3-(Boc-amino)-3-(4-methoxyphenyl)propionsäure
3-(Boc-amino)-3-(4-methoxyphenyl)propionsäure
Formel:
C₁₅H₂₁NO₄ Molecular Weight: 295.3309 g/mol |
MDL:
MFCD02090694 CAS-Nummer: 96363-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...