(S)-(+)-2-Fluor-α-methyl-4-biphenylessigsäure

Lieferant: Cayman Chemical

Synonyme: (S)-Flurbiprofen, (S)-2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid

10004207-100 10004207-500
CAYM10004207-100EA 129 EUR
CAYM10004207-100 CAYM10004207-500
(S)-(+)-2-Fluor-α-methyl-4-biphenylessigsäure
(S)-(+)-2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
Molecular Weight: 244,260 g/mol
MDL: MFCD00866152
CAS-Nummer: 51543-39-6
EINECS: 257-263-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR