trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: (E)-4,4,5,5-Tetramethyl-2-(3-((tetrahydro-2H-pyran-2-yl)oxy)prop-1-en-1-yl)-1,3,2-dioxaborolane

PN-2529-1G PN-2529-250M
COBBPN-2529-1GEA 595 EUR
COBBPN-2529-1G COBBPN-2529-250M
trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronsäurepinakolester
trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronsäurepinakolester
Formel: C₁₄H₂₅BO₄
Molecular Weight: 268,16 g/mol
MDL: MFCD03788736
CAS-Nummer: 642066-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR