3-(Boc-amino)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl (3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, 3-(Boc-amino)benzeneboronic acid pinacol ester

PN-2621-5G PN-2621-1G
COBBPN-2621-5GEA 374 EUR
COBBPN-2621-5G COBBPN-2621-1G
3-(Boc-amino)phenylboronsäurepinakolester
3-(Boc-amino)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₄
Molecular Weight: 319,21 g/mol
MDL: MFCD03789256
CAS-Nummer: 330793-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR