3-(Boc-amino)phenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
tert-Butyl (3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, 3-(Boc-amino)benzeneboronic acid pinacol ester
010780-5G
010780-1G
FLUO010780-5GEA
567
EUR
FLUO010780-5G
FLUO010780-1G
3-(Boc-amino)phenylboronsäurepinakolester
3-(Boc-amino)phenylboronsäurepinakolester
Formel:
C₁₇H₂₆BNO₄ Molecular Weight: 319.21 g/mol |
MDL:
MFCD03789256 CAS-Nummer: 330793-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...