3-Hydroxy-5-(methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Hydroxy-5-(methoxycarbonyl)phenylboronic acid, pinacol ester, Methyl 3-hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

PN-5977-1G PN-5977-250M
COBBPN-5977-1GEA 1040 EUR
COBBPN-5977-1G COBBPN-5977-250M
3-Hydroxy-5-(methoxycarbonyl)phenylboronsäurepinakolester
3-Hydroxy-5-(methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₅
Molecular Weight: 278.108 g/mol
MDL: MFCD12546597
CAS-Nummer: 1004294-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR