3-(2-Cyanethylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-((2-Cyanoethyl)carbamoyl)phenyl)boronic acid, 3-(2-Cyanoethylaminocarbonyl)benzeneboronic acid

BB-3066-5G BB-3066-1G
COBBBB-3066-5GEA 776 EUR
COBBBB-3066-5G COBBBB-3066-1G
3-(2-Cyanethylcarbamoyl)phenylboronsäure
3-(2-Cyanethylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₁BN₂O₃
Molecular Weight: 218.0200 g/mol
MDL: MFCD04115697
CAS-Nummer: 762262-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR