3-(Boc-amino)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Aminobenzeneboronic acid, N-BOC protected, 3-(N-Boc-amino)phenylboronic acid, (3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid, 3-Boronoaniline, N-BOC protected, 3-[(tert-Butoxycarbonyl)amino]benzeneboronic acid, 3-(Boc-amino)benzeneboronic acid

BB-2621-1G BB-2621-5G BB-2621-25G
COBBBB-2621-1GEA 0 EUR
COBBBB-2621-1G COBBBB-2621-5G COBBBB-2621-25G
3-(Boc-amino)phenylboronsäure
3-(Boc-amino)phenylboronsäure
Formel: C₁₁H₁₆BNO₄
Molecular Weight: 237.06 g/mol
MDL: MFCD03411945
CAS-Nummer: 380430-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR