2-Amino-4-(isopropoxycarbonyl)phenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

BB-5724-1G BB-5724-5G
COBBBB-5724-1GEA 303 EUR
COBBBB-5724-1G COBBBB-5724-5G
2-Amino-4-(isopropoxycarbonyl)phenylboronsäure Hydrochlorid
2-Amino-4-(isopropoxycarbonyl)phenylboronsäure Hydrochlorid
Formel: C₁₀H₁₅BClNO₄
MDL: MFCD12025975
CAS-Nummer: 1150114-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR