3,5-Bis(methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Bis(methoxycarbonyl)benzeneboronic acid, (3,5-Bis(methoxycarbonyl)phenyl)boronic acid

BB-5280-5G BB-5280-1G
COBBBB-5280-5GEA 813 EUR
COBBBB-5280-5G COBBBB-5280-1G
3,5-Bis(methoxycarbonyl)phenylboronsäure
3,5-Bis(methoxycarbonyl)phenylboronsäure
Formel: C₁₀H₁₁BO₆
Molecular Weight: 238.00 g/mol
MDL: MFCD11053854
CAS-Nummer: 177735-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR