3-(tert-Butyldimethylsilyloxy)-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(t-Butyldimethylsilyloxy)-4-methoxyphenylboronic acid

BB-5650-1G BB-5650-5G
COBBBB-5650-1GEA 310 EUR
COBBBB-5650-1G COBBBB-5650-5G
3-(tert-Butyldimethylsilyloxy)-4-methoxyphenylboronsäure
3-(tert-Butyldimethylsilyloxy)-4-methoxyphenylboronsäure
Formel: C₁₃H₂₃BO₄Si
Molecular Weight: 282,2200 g/mol
MDL: MFCD11855851
CAS-Nummer: 900152-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR