4-(tert-Butyldimethylsilyloxy)-3-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3837-5G BB-3837-1G
COBBBB-3837-5GEA 1180 EUR
COBBBB-3837-5G COBBBB-3837-1G
4-(tert-Butyldimethylsilyloxy)-3-methoxyphenylboronsäure
4-(tert-Butyldimethylsilyloxy)-3-methoxyphenylboronsäure
Formel: C₁₃H₂₃BO₄Si
MDL: MFCD09027250
CAS-Nummer: 957060-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR