4-Methoxy-3-(piperidin-1-ylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methoxy-3-(piperidin-1-ylsulphonyl)phenyl)boronic acid

BB-2919-5G BB-2919-1G
COBBBB-2919-5GEA 1690 EUR
COBBBB-2919-5G COBBBB-2919-1G
4-Methoxy-3-(piperidin-1-ylsulfonyl)phenylboronsäure
4-Methoxy-3-(piperidin-1-ylsulfonyl)phenylboronsäure
Formel: C₁₂H₁₈BNO₅S
Molecular Weight: 299.1500 g/mol
MDL: MFCD08235037
CAS-Nummer: 874219-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR