4-(Tetrahydropyran-2-yloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(2-Tetrahydropyranyloxy)benzeneboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)phenylboronic acid, 4-(Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, (4-((Tetrahydro-2H-pyran-2-yl)oxy)phenyl)boronic acid

BB-2678-5G BB-2678-25G
COBBBB-2678-5GEA 56 EUR
COBBBB-2678-5G COBBBB-2678-25G
4-(Tetrahydropyran-2-yloxy)phenylboronsäure
4-(Tetrahydropyran-2-yloxy)phenylboronsäure
Formel: C₁₁H₁₅BO₄
Molecular Weight: 222,05 g/mol
MDL: MFCD02183525
CAS-Nummer: 182281-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR