3-(tert-Butyldimethylsilyloxy)phenylboronsäure ≥90%, tech.

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(tert-Butyl dimethylsiloxy)phenyl boronic acid, (3-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid, 3-(tert-Butyldimethylsiloxy)benzeneboronic acid

H28378.03 H28378.06
H28378.03EA 57.2 EUR
H28378.03 H28378.06
3-(tert-Butyldimethylsilyloxy)phenylboronsäure ≥90%, tech.
3-(tert-Butyldimethylsilyloxy)phenylboronsäure

Purity: 90% CAS No.: 261621-12-9 Molecular Formula: C12H21BO3Si Formula Weight: 252.19 Melting Point: 86-91° MDL No.: MFCD03701505

Formel: C₁₂H₂₁BO₃Si
Molecular Weight: 252.19 g/mol
Schmelzpunkt: 86…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03701505
CAS-Nummer: 261621-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR