4-(tert-Butyldimethylsilyloxy)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(tert-Butyl dimethylsiloxy)phenyl boronic acid, 4-(tert-Butyldimethylsiloxy)phenyl boronic acid

M-Clarity™ Qualitätsstufe = MQ100

524042-1G 524042-5G
SIAL524042-1GEA 65.2 EUR
SIAL524042-1G SIAL524042-5G
4-(tert-Butyldimethylsilyloxy)phenylboronsäure, Sigma-Aldrich®
4-(tert-Butyldimethylsilyloxy)phenylboronsäure
Formel: C₁₂H₂₁BO₃Si
Molecular Weight: 252,1900 g/mol
MDL: MFCD03093888
CAS-Nummer: 159191-56-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR