3-(Piperidin-1-ylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Piperidin-1-ylsulphonyl)phenyl)boronic acid

BB-2349-5G BB-2349-1G
COBBBB-2349-5GEA 1880 EUR
COBBBB-2349-5G COBBBB-2349-1G
3-(Piperidin-1-ylsulfonyl)phenylboronsäure
3-(Piperidin-1-ylsulfonyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₄S
Molecular Weight: 269,1300 g/mol
MDL: MFCD07783850
CAS-Nummer: 690662-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR