4-(Piperidin-1-ylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Piperidin-1-ylsulphonyl)phenyl)boronic acid

BB-2425-5G BB-2425-1G
COBBBB-2425-5GEA 796 EUR
COBBBB-2425-5G COBBBB-2425-1G
4-(Piperidin-1-ylsulfonyl)phenylboronsäure
4-(Piperidin-1-ylsulfonyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₄S
Molecular Weight: 269,1250 g/mol
MDL: MFCD06659850
CAS-Nummer: 486422-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR