3-(Morpholinosulfonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-(Morpholinosulphonyl)phenyl)boronic acid, 3-(4-Morpholinylsulphonyl)benzeneboronic acid
BB-2351-5G
BB-2351-1G
COBBBB-2351-5GEA
914
EUR
COBBBB-2351-5G
COBBBB-2351-1G
3-(Morpholinosulfonyl)phenylboronsäure
3-(Morpholinosulfonyl)phenylboronsäure
Formel:
C₁₀H₁₄BNO₅S Molecular Weight: 271.09 g/mol |
MDL:
MFCD07363748 CAS-Nummer: 871329-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...