4-Brom-2-chlor-5-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromo-2-chloro-5-methoxybenzeneboronic acid
BB-2596-5G
BB-2596-1G
COBBBB-2596-5GEA
884
EUR
COBBBB-2596-5G
COBBBB-2596-1G
4-Brom-2-chlor-5-methoxyphenylboronsäure
4-Brom-2-chlor-5-methoxyphenylboronsäure
Formel:
C₇H₇BBrClO₃ Molecular Weight: 265.30 g/mol |
MDL:
MFCD06659862 CAS-Nummer: 850567-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...