6-Brom-2-chlor-3-methoxyphenol
Lieferant: COMBI-BLOCKS
OT-1225-1G
OT-1225-5G
COBBOT-1225-1GEA
303
EUR
COBBOT-1225-1G
COBBOT-1225-5G
6-Brom-2-chlor-3-methoxyphenol
6-Brom-2-chlor-3-methoxyphenol
Formel:
C₇H₆BrClO₂ |
MDL:
MFCD16618957 CAS-Nummer: 1228957-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...