5-Brom-2-fluor-N-methylanilin
Lieferant: COMBI-BLOCKS
HC-2872-5G
HC-2872-1G
COBBHC-2872-5GEA
1040
EUR
COBBHC-2872-5G
COBBHC-2872-1G
5-Brom-2-fluor-N-methylanilin
5-Brom-2-fluor-N-methylanilin
Formel:
C₇H₇BrFN |
MDL:
MFCD09745011 CAS-Nummer: 1153252-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...