4-Brom-2-fluor-N-methylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-3277-5G HC-3277-1G
COBBHC-3277-5GEA 1110 EUR
COBBHC-3277-5G COBBHC-3277-1G
4-Brom-2-fluor-N-methylanilin Hydrochlorid
4-Brom-2-fluor-N-methylanilin Hydrochlorid
Formel: C₇H₈BrClFN
MDL: MFCD12913131
CAS-Nummer: 1197930-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR